Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Supplier: VWR
VWR® Talon®
915748 915746 915745 915744
14224-402EA 1638.79 USD InStock
14224-402 14224-400 14224-406 14224-404
VWR® Talon® Stainless Steel Fume Hood Lab-Frame Kits
Support Rods and Frames
Choose from four kits specifically designed to fit within laboratory fume hoods.
  • Stainless steel construction of rods
  • Nickel-plated zinc connectors and lab frame feet
  • Variety of kits designed to fit within laboratory fume hoods
The 1.5m (5') kit includes four 127 cm (50") horizontal rods, five 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 34 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 1.2m (4') kit includes four 96.5 cm (38") horizontal rods, four 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 30 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 1.8m (6') kit includes four 157.5 cm (62") horizontal rods, six 91.4 cm (36") vertical rods, two 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, eight 5.1 cm (2") rods, 38 lattice connectors, four rod-end connectors, and four Lab Frame feet.
The 2.4m (8') kit includes four 218.5 cm (86") horizontal rods, eight 91.4 cm (36") vertical rods, three 30.5 cm (12") base rods, two 45.7 cm (18") rear support rods, ten 5.1 cm (2") rods, 45 lattice connectors, four rod-end connectors, and six Lab Frame feet.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty