Would you like to log in again?
Mini-Unions and Ferrules, Graphite and Graphite/Vespel®, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
072695 072629 103431 103430 072696 072630 072698 072697 103433 072694 103432
14226-558EA 68.17 USD InStock
14226-558 14226-556 14226-348 14226-554 14226-356 14226-552 14226-354 14226-352 14226-350 14226-282 14226-280
Mini-Unions and Ferrules, Graphite and Graphite/Vespel®, Trajan Scientific and Medical
Fittings and Connectors
These unions are used for fitting a pre-column (guard column) and for GC x GC connections.
  • Specifically designed for connecting capillary columns
  • Easy way of connecting guard columns or retention gaps to capillary columns
  • Can be used for repairing broken columns
  • Inert and precise glass lined bore
  • Supplied with 15% Graphite, 85% Vespel® ferrules
Your chromatography analysis does not end with the selection of the GC column. The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes. SGE mini-Unions are specifically designed and precision engineered for use in capillary GC.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: Mini-Unions and replacement ferrules are available in sizes to connect 0.1 - 0.25 mm I.D., 0.32 mm I.D. and 0.53 mm I.D. columns and fused silica tubing. 100% graphite ferrules available separately. Packs contain 1 mini union, 2 nuts and 5 ferrules.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty