Would you like to log in again?
SilTite® Metal Ferrules, SGE
Supplier: Trajan Scientific and Medical
SilTite™
073224 073225
14226-386PK 100.74 USD InStock
14226-386 14226-384
SilTite® Metal Ferrules, SGE
Fittings and Connectors
SilTite metal ferrules have been developed specifically to overcome the problems associated with the use of 100% graphite and composite ferrules
  • Strong seal on capillary columns
  • Reduced air background
  • No contamination
  • Cleaner spectra
  • Leak free
  • Nut does not need re-tightening after initial temperature cycles
SilTite ferrules form a strong and permanent airtight seal around the capillary column and have a temperature limit well above the temperature capacity of the injector, MS interface or GC oven. The metal SilTite ferrule together with the metal nut have the same coefficient of thermal expansion. The ferrule and nut expand and contract at the same rate eliminating any chance of annoying leaks. The nut does not need to be re-tightened after initial temperature cycles.
Certifications: RoHS compliant per Directive 2011/65/EU
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty