Would you like to log in again?
Trajan, SilTite™ Mini-Unions, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
SilTite™
073551 073552 073470 073550 073471 073472 073553
14226-438EA 147.84 USD InStock
14226-438 14226-436 14226-434 14226-442 14226-432 14226-440 14226-430
Trajan, SilTite™ Mini-Unions, Trajan Scientific and Medical
Fittings and Connectors
Your chromatography analysis does not end with the selection of the GC column.
  • Inert and precise glass lined bore
  • Robust and stable
  • For both extreme temperature and pressure cycling
  • Leak-free
  • No need to re-tighten after installation
  • Low dead volume
These unions should be used when fitting a pre-column or guard column, GC to GC connections. They can be used to repair broken columns.
The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes. SGE SilTite™ mini-Unions are specifically designed and precision engineered for use in capillary GC.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: SilTite™ mini-Unions and replacement ferrules are available in sizes to connect 0.1 - 0.25mm ID, 0.32 mm I.D. and 0.53 mm I.D. columns and fused silica tubing. Packs contain 1 mini union, 2 nuts and 5 ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty