Would you like to log in again?
Ovation® Fixed Volume Pipettors, VistaLab
Supplier: VistaLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Ovation
1257-0250 1257-1000 1257-0001 1157-0025 1157-0010 1257-0002 1257-0010 1257-0020 1157-0500 1157-0200 1257-0050 1157-0250 1257-0200 1157-0020 1257-0300 1157-0050 1157-1000 1257-0500 1257-0005 1157-0100 1157-0999 1257-0100 1157-0300 1257-0025 1157-0005
14227-434EA 438.39 USD InStock
14227-434 75814-372 14227-436 14227-458 14227-452 99089-398 14227-432 14227-454 14227-450 14227-428 14227-468 14227-424 14227-446 14227-426 14227-448 14227-464 14227-442 14227-444 14227-466 14227-422 14227-460 14227-462 14227-440 75814-376 14227-438
Ovation® Fixed Volume Pipettors, VistaLab
Pipettes Repetitive Pipettors
Mechanical 1-stroke (F1) or 2-stroke (F2) pipettors with low-force plunger operation.
  • Ovation F1 models available in 11 fixed volumes ranging from 5 to 1000 μl
  • Ovation F2 models available in 13 fixed volumes ranging from 1 to 1000 μl
  • Built-in microtube cap opener
  • Custom fixed volumes available upon request
Ovation pipettors and essential components are available, with pipettors featuring mechanical low-pressure plunger operation.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty