Would you like to log in again?
Seals for Headspace Vials, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W224217 W224225 W224221 W224223 W224224 W224216 W224215
14227-786CS 272.03 USD InStock
14227-786 14227-788 89495-262 89495-258 89495-256 14227-784 89495-260
Seals for Headspace Vials, Wheaton, DWK Life Sciences
Closures Chromatography Vial Closures
Pressure release seals feature score lines that allow for internal pressure release when 3.0 ±0.5 bar has been exceeded.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty