Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HD-ESD System Coveralls, WORKLON®

Supplier: Superior Uniform Group

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Worklon®
1956-S 1956-5XL 1956-XS 1956-2XL 1956-4XL 1956-L 1956-XL 1956-M 1956-3XL
14227-930EA 284.84 USD InStock
14227-930 14227-932 14227-926 14227-938 14227-928 14227-922 14227-934 14227-924 14227-936
HD-ESD System Coveralls, WORKLON®
Workwear
Reusable static dissipative coveralls are designed for ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom environments, where bacterial filtration, static charge, particulate resistance, and fluid resistance control is required

They feature raglan sleeves, a zipper front closure, a two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar and ankles. Three vertical snaps on the back of the legs accommodate an adjustable boot attachment. A color-coded hang loop denotes size. Gamma compatible. Autoclavable.

Coveralls are constructed of 99% polyester with 1% carbon yarn stripe fabric.

Ordering information: Boots are available. Contact your Avantor representative for information.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty