Would you like to log in again?
SC-3 System Coveralls, WORKLON®
Supplier: Superior Uniform Group
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Worklon®
1910-2XL 1910-5XL 1910-M 1910-XS 1911-XL 1911-XS 1911-3XL 1911-5XL 1910-L 1911-S 1911-M 1910-XL 1911-4XL 1910-3XL 1910-S 1911-2XL 1911-L 1910-4XL
14227-940EA 433.81 USD InStock
14227-940 14227-962 14227-974 14227-952 14227-942 14227-964 14227-954 14227-970 14227-960 14227-950 14227-972 14227-948 14227-944 14227-966 14227-956 14227-946 14227-968 14227-958
SC-3 System Coveralls, WORKLON®
Workwear
Reusable coveralls are designed for use in microelectronics, semiconductor, disk drive, laser, and other industrial ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanrooms
Coveralls have raglan sleeves, a zipper front with two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar and ankles. Three vertical snaps on the back of the legs accommodate an adjustable boot attachment. For boots, see 14227-884 series. A color-coded hang loop denotes size.
The Burlington C3™ high density fabric dissipates static charges and guards against airborne particles and lint.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty