Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SC-3 System Frocks, WORKLON®
Supplier: Superior Uniform Group
Worklon®
3440-4XL 3442-2XL 3442-M 3441-2XL 3441-XL 3440-2XL 3442-5XL 3441-3XL 3442-4XL 3440-S 3440-M 3441-S 3440-XL 3441-XS 3440-5XL 3440-3XL 3441-4XL 3442-L 3442-3XL 3442-XL 3440-XS 3441-5XL 3442-XS 3441-L 3440-L 3442-S
14227-996EA 359.55 USD InStock
14227-996 14227-998 14227-976 14227-992 14227-994 14227-990 14228-024 14228-002 14228-022 14228-000 14228-028 14228-006 14228-026 14228-004 14227-988 14228-020 14227-984 14227-986 14227-982 14228-008 14228-014 14228-012 14228-018 14228-016 14228-010 14227-978
SC-3 System Frocks, WORKLON®
Workwear
Reusable frocks are designed for use in microelectronics, semiconductor, disk drive, laser, and other ISO Class 6 (FED-STD-209E Class 1000/M4.5) industrial cleanrooms.
  • Available with snap front closure or zipper front closure
  • Color-coded hang loop denotes size
The Burlington C3™ high density fabric dissipates static charges and guards against airborne particles and lint. Reusable cleanroom frocks have a two-ply reinforced placket, antistatic knit cuffs, and adjustable snaps at the collar.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty