Would you like to log in again?
Revco® Accessories for Refrigerators and Freezers, Thermo Scientific
Supplier: Thermo Scientific
Revco®
6701 6202 6704 6183-7 6383-7A 6383-7D 90520H07 6702 4425 6703 90520H11 6206 6224TA 5612-1A 4706 6700 6183-7A 6736 6041 6105 6903 4426 6383-6A 6694 6383-5
55702-641PK 191.21 USD InStock
55702-641 89176-742 14228-318 89176-744 55701-740 73620-026 73650-100 CA14228-372 55702-851 14228-308 89176-758 CA89369-376 14228-310 14228-255 14228-376 14228-352 14228-374 14228-314 14228-512 14228-378 14228-312 14228-356 73620-050 14228-372 CA55702-630
Revco® Accessories for Refrigerators and Freezers, Thermo Scientific
Refrigerators
Temperature recorders use pressure-sensitive chart paper (sold separately) and do not require inking
  • A built-in surge suppressor is standard on Ultima II series models
  • A freestanding surge suppressor is available for Elite® and Value series models
  • Revco® ultra-low temperature freezers include one 2.5 cm (1") diameter pass-thru port
  • Additional ports are available to provide openings for probes, cables, or leads
The remote alarm module in temperature recorder provides an audible and visual signal in the event of a temperature rise or power failure
Built-in backup systems are available on Ultima® II and Elite® models only; Value series models require a freestanding backup system. All Ultima® II, Elite®, and Value series 88L (3.1cu.ft.) –86°C models require a freestanding backup system as well.
A built-in surge suppressor is standard on Ultima II series models. A freestanding surge suppressor is available for Elite® and Value series models. Revco® ultra-low temperature freezers include one 2.5 cm (1") diameter pass-thru port. Additional ports are available to provide openings for probes, cables, or leads.
The deluxe electronic remote alarm can be programmed to sound an alarm in the event of a temperature increase or power failure. It is able to dial up to four telephone numbers and can monitor up to four individual freezers or up to three groups of freezers.
Ordering information: Many accessories listed are factory-installed and must be specified when ordering. The remote alarm module does not include a wire. The deluxe electronic remote alarm requires a dedicated phone line.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty