Would you like to log in again?
MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Supplier: Sorenson BioScience
MµltiFit™
31900 10590 10040 15670 32100 32000 10660 15370 10190T 15670T 10520 15720T 10190 15730 30150T 30140T 10200T 15720 10130 30430T 10080 10470 10230 10230T 10470T 10620 10520T 30440 31800T 15270T 10200 10500 15660T 30450T
53550-103CS 484.97 USD InStock
53550-103 14229-740 14229-742 14229-886 53550-123 53550-101 53550-087 53550-065 14229-832 14229-876 14229-892 53550-113 14229-750 14229-894 14229-872 14229-796 14229-830 14229-874 53550-096 53550-094 53550-099 53550-076 53550-093 53550-090 14229-744 14229-888 14229-866 14229-822 14229-746 14229-824 14229-868 14229-748 14229-826 14229-828
MµltiFit™ Universal Fit Pipette Tips, Sorenson BioScience
Pipette Tips
Pipette tips feature a unique beveled end to minimize fluid retention.
  • Fully autoclavable
  • Tips fit most popular research-grade pipettors
They ensure superior sample delivery and recovery in critical micro-volume ranges. Reinforced base design and standard 96-well pattern of cell racks ensures rapid mounting of small tips on all multichannel pipettors.
Certifications: The 1 to 200 µl racked tips are certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in bulk packaging, autoclavable polypropylene racks, hinged racks, QuickRack reloads for reloading standard racks, and QuickShot reloads for reloading hinged racks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty