Would you like to log in again?
MµltiFit™ Micro Volume Pipette Tips, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MµltiFit™
30470T 23560T 37640T 37640 13980 37650T 22210 37660 37650 23580T 13980T 37820T 22220-E 23570T
53550-006CS 416.81 USD InStock
53550-006 53550-004 14229-860 14229-862 14229-764 14229-930 53550-001 14229-928 14229-932 14229-878 14229-858 14229-924 14229-926 14229-938
MµltiFit™ Micro Volume Pipette Tips, Sorenson BioScience
Pipette Tips
These natural-colored tips fit most popular brand research-grade pipettors.
They feature a narrow bore that reduces dead air space around extended air-displacement piston. Narrow bore also increases accuracy and reproducibility for critical ultra microvolume ranges.
Certifications: Certified RNase-, DNase-, human DNA-, PCR inhibitor-, and pyrogen-free.
Ordering information: Available in autoclavable polypropylene racks, bulk packed, or QuickRack reloads for the reloading racks.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty