Would you like to log in again?
Seracult® Fecal Occult Blood Tests, Propper Manufacturing
Supplier: Propper
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Seracult®
37901500 37200500 37901600 37100100 37901000 37100200 37200300 37901700 37200400
10753-456CS 660.1 USD InStock
10753-456 14230-050 60829-035 10753-472 14230-049 14230-027 10753-470 14230-039 14230-048
Seracult® Fecal Occult Blood Tests, Propper Manufacturing
Clinical Diagnostic Supplies Clinical Reagents and Kits
Seracult® fecal occult blood test is a qualitative aid in the diagnosis of colorectal cancer.
  • Guaiac fecal occult blood test to aid in the detection of colorectal cancer
  • Single or Triple Slide
  • Long Sample Stability
  • Fast qualitative results
  • Mailing kits of in-office slides
  • 28 days sample stability
Seracult® Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer. Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home. Seracult® is available in a variety of configurations to meet all of your needs. Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty