Would you like to log in again?
Nonsterile Transport Tubes, 10 ml, Simport Scientific
Supplier: Simport
T550WOS T550W T550-10AT T550-10ATPR T550W T550-10AT T550WOS T550-10ATPR
14231-890CS 426.36 USD InStock
CA60872-810 CA60872-812 14231-890 CA60872-811 14231-888 14231-884 CA60872-809 14231-886
Nonsterile Transport Tubes, 10 ml, Simport Scientific
Tubes Test Tubes
Designed for storage and transportation of biological material.
  • Manufactured from nontoxic polypropylene
  • Tubes have external threads and a smooth, uniform inner surface that reduces the risk of contamination
  • These virtually unbreakable tubes feature a self-standing base and clear sides
Tubes 14231-886 feature graduations from 1 to 9 ml in 0.5 ml increments and a white marking area for sample identification. Polyethylene caps 14231-888 feature a flexible inner sealing lip that ensures a leakproof seal. Polyethylene caps 14231-890 feature a silicone O-ring for the most secure seal. All caps also feature a long skirt and may be removed or sealed with only a single turn.
Ordering information: Tubes and caps sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty