Would you like to log in again?
Filter Tips, Sun Sri
Supplier: SUN SRi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
204 004 204 002
14232-306CS 458.35 USD InStock
14232-306 14232-308
Filter Tips, Sun Sri
Filters Filter Tubes
These dissolution filters are manufactured of polyethylene with a limited porous area.
Fits securely on the end of a ¹/₈" cannula or sample probe.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty