Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titan3™ Syringe Filters, 30 mm, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Titan3™
41225-GMC 44525-PC 42225-NC 44525-NP 44525-PV
14232-428CS 1548.85 USD InStock
14232-428 14235-360 76111-608 14232-324 14232-754
Titan3™ Syringe Filters, 30 mm, Thermo Scientific
Filters Syringe Filters
Banded polypropylene housing ensures lab safety, preventing the housing from bursting.

  • Color-coded housing allows for easy identification of membrane
  • Most 30 mm filters have a 1 µm glass pre-filter
  • For sample volumes of 2 to 100 ml
  • Retention volume <115 µl
  • Maximum operating temperature: 100 °C (autoclavable at 125 °C for 15 minutes)
  • Maximum operating pressure is approximately 100 psi

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty