Would you like to log in again?
Plastic Disposable Syringes, SUN SRi
Supplier: SUN SRi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
501 370 400027 400 010 400 018 400 030 400 021 501 369 400 008 400 012 400 014 201644 400 029 400 004
14232-280CS 755.93 USD InStock
14232-280 14235-480 14235-526 14235-407 14235-512 14232-898 14232-514 14232-298 14232-486 14232-398 14232-396 14232-278 14232-300
Plastic Disposable Syringes, SUN SRi
Syringes General-Purpose Syringes
Disposable syringes made of inert polypropylene and polyethylene.
  • Chemically resistant
  • Easy to read permanent graduations
  • Safe functional back-stop feature
No rubber plunger seals or silicone lubricants that may cause sample contamination.
Packaging: Bulk packaged.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty