Would you like to log in again?
Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Supplier: Metro International
Super Erecta® Super Erecta Shelf®
AW31C SW45C 2WS14S SW31C 63PDF SW23C 13PDF 1WD14S 1WD24C SW21C SW55C 1WD14S AW35C 2WS21C 1WS21C SW43C SW56C 13PDF 1WD24C BCS 1WS18C AW56C 2WD24S SW53C 1WS14C 2WD18S 1WS21C SW43C AW35C 1WS14S 2WS18S BES 2WD24C 2WD21C 2WS24S 1WS24S AW45C AW41C AW33C AW45C 33PDF SW41C AW41C 2WD21C 1WD24S SW31C 1WS21S 33PDFS AW25C AW36C 1WD24S 63PDFS 1WS18S AW46C SBCS 2WD18C SW56C AW36C SW51C SW35C 1WD14C SW25C 2WS14C 2WS18S 2WS21C 2WD24S AW23C 1WS21S 2WS18C 2WD24C 13PDFS SW35C SW51C 2WS24C SW36C 1WS24C 2WS24C SW25C 1WS14C 1WD18C 2WD21S AW33C AW23C AW21C AW51C 2WD14C 2WS18C AW55C 1WS18S SW36C AW25C 1WD18S AW53C 63PDFS 54PDFS BCS AW51C 1WD18S 63PDF 13PDFS AW26C 1WS24C 1WD21C BES 1WS14S 1WD21S SW23C AW46C 2WD14S SW33C 2WS14S AW21C AW26C 2WS14C AW43C AW55C 2WD21S 2WD18C SW41C 2WS21S 2WD18S 1WD14C AW53C 2WS21S 1WD21S SW26C 1WS18C 1WD18C 33PDF 2WD14C SW26C SW45C AW31C SW55C 54PDF SW46C SW53C 54PDF SBCS 2WS24S 54PDFS AW56C AW43C SW21C 2WD14S 33PDFS SW46C 1WS24S 1WD21C SW33C
14233-028EA 402.89 USD InStock
14233-028 CA14233-038 CA14233-158 14233-020 CA14233-030 14233-142 CA14233-150 CA14233-032 14233-022 14233-144 CA14233-152 14233-024 CA14233-034 14233-146 CA14233-154 14233-026 CA14233-036 CA14233-156 14233-148 14233-140 14233-138 CA14233-148 CA14233-026 14233-018 CA14233-028 14233-130 CA14233-140 14233-098 CA14233-142 14233-132 CA14233-020 CA14233-144 14233-134 CA14233-022 CA14233-146 14233-136 CA14233-024 14233-016 14233-090 14233-092 14233-094 14233-096 CA14233-052 14233-042 14233-164 14233-044 CA14233-054 14233-166 CA14233-056 14233-046 14233-048 CA14233-058 14233-160 14233-040 CA14233-050 14233-162 CA14233-048 CA14233-162 14233-152 14233-032 CA14233-040 14233-154 CA14233-164 CA14233-042 14233-034 CA14233-166 14233-156 CA14233-044 14233-036 14233-158 CA14233-046 14233-038 14233-150 CA14233-160 14233-030 14233-106 CA14233-114 14233-108 CA14233-116 CA14233-118 14233-064 CA14233-074 14233-066 CA14233-076 14233-068 CA14233-078 14233-102 CA14233-110 14233-104 CA14233-112 14233-060 CA14233-070 CA14233-072 14233-062 CA14233-104 CA14233-106 CA14233-108 CA14233-062 14233-054 14233-056 CA14233-064 14233-058 CA14233-066 CA14233-102 CA14233-068 14233-050 CA14233-060 14233-052 CA14233-016 14233-128 CA14233-136 CA14233-018 CA14233-138 CA14233-096 14233-086 14233-120 14233-088 CA14233-098 CA14233-130 14233-122 14233-124 CA14233-132 14233-126 CA14233-134 14233-080 CA14233-090 CA14233-092 14233-082 14233-084 CA14233-094 14233-116 CA14233-126 14233-118 CA14233-128 14233-076 CA14233-084 14233-110 CA14233-120 CA14233-086 14233-078 14233-112 CA14233-122 CA14233-088 14233-114 CA14233-124 14233-070 CA14233-080 14233-072 CA14233-082 14233-074
Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Furniture Shelves
Wall mounts for shelves are an ideal method for storing items overhead or above work surfaces.
  • All prepackaged post-type units feature a chrome-plated finish
  • Metal endcaps to cover post openings are provided
  • Use single supports at shelf ends and double supports for adjoining shelves
  • Direct wall mounts are available in chrome-plated or stainless steel finish
  • Direct wall systems have a maximum load rating of 91 kg (200 lbs.)
  • Post-type systems have a maximum load rating of 113.3 kg (250 lbs.)
Post-type wall mounts are available in prepackaged end units or mid units, which include all necessary components except wall bolts or screws. End units consist of two posts with brackets, and two single shelf supports for each shelf. Mid units consist of one post with brackets and one double shelf support for each tier. If shelving is to be the length of one shelf, order one end unit only.
Posts, brackets, and shelf supports may also be purchased separately to construct post-type wall mount systems. Each post is supplied with two end brackets. The 85.4 cm (33⁵/₈") long posts also include one intermediate bracket; the 137.2 cm (54") and 157.5 cm (62") long posts also include two intermediate brackets. Intermediate brackets can also be used to attach freestanding shelving to a wall. Post-type components are available in chrome-plated or stainless steel finish.
Direct wall mounts eliminate the need for a post; the shelf support is held by a mounting plate attached directly to the wall. This design also conserves space, as it allows the shelf and shelf support to be much closer to the wall. Each direct wall mount consists of one shelf support, one bracket, and two metal endcaps.
Brackets accommodate all types and sizes of Super Erecta® shelves (shelves sold separately).
Ordering information: If shelving is to be the length of two shelves, order one end unit and one mid unit; if three shelf lengths, order one end and two mid units and so on.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty