Would you like to log in again?
Rubber Work Overshoes, Tingley
Supplier: Tingley
1400M 1400L 1400XL 1400S 1400XXXL 1400XXL
14233-315CS 850.89 USD InStock
14233-315 83007-898 14233-317 15001-833 83007-901 14233-313
Rubber Work Overshoes, Tingley
Shoes and Boots
100% waterproof overshoes made from a injection-molded, seamless rubber construction and a deep cleated outsole, which releases debris and provide good slip resistance. The boots stretch easily to fit even over the bulkiest workshoes.
  • Reinforced heel and toe
  • Excellent tensile strength
  • Heavy-duty heel kick-off cleat for easy no-hands removal
  • Molded-in button for secure closure
  • Material will not crack or stiffen in cold weather
  • Unlined for easy cleaning and decontamination
  • Additional material in the heel, toe, and sole provide longer wear; less material in the upper reduces weight and leg fatigue
Designed to resist acids, alcohols, bases, and diluted water solutions of most water-soluble chemicals.
Color: Black
Height: 25.4 cm (10")
Caution: Not recommended for constant contact with petroleum and oil-based products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty