Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Finntip™ Wide Orifice Pipette Tips, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Finntip™
9405163 9405160 9405050 9405020
53516-240PK 129.73 USD InStock
53516-240 14233-398 53516-244 14233-400
Finntip™ Wide Orifice Pipette Tips, Thermo Scientific
Pipette Tips
Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips.

  • Two different models cover volume range of 10 - 1000 μL
  • Wide orifice tip; inside diameter of orifice is 1.1 mm/1.8 mm
  • Color code: White
  • Length: 5.3 cm/7.2 cm

Ideal for pipetting macromolecules.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty