Would you like to log in again?
MµltiGuard™ Barrier Pipette Tips, Sterile, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MµltiGuard™
36060 14210T 14200 38000T 14220T 14210 14200T 15020 15020T 14220 28200T 28200
14234-114CS 833.72 USD InStock
14234-114 14234-102 14234-112 14234-122 14234-100 14234-120 14234-098 14234-094 14234-108 14234-092 14234-106 14234-104
MµltiGuard™ Barrier Pipette Tips, Sterile, Sorenson BioScience
Pipette Tips
Ideal for PCR, radioisotope labeling, and pipetting procedures that require an aerosol-free method for handling samples.
  • Tips fit most popular research-grade pipettors
Low-binding tips feature reduced surface tension which helps minimize the bonding of DNA and protein molecules to the tip material.
Ordering information: Supplied in polypropylene CellRacks. MµltiGuard™ tips 14234-104 and 14234-106 can accommodate a volume of 30 µl.
Packaging: Packs contain 10 racks with 96 tips/rack, except 14234-120 and 14234-122, which have 100 tips/rack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty