Would you like to log in again?
Boston Round Bottles, Clear, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WHEATON®
W216803 W216810 W216830 W216804 W216815 W216824 W216821 W216823 W216834 W216833 W216827 W216801 W216805 W216809 W216818 W216800 W216831 W216822 W216811 W216814 W216825 W216832 W216829 W216808 W216806 W216802 W216813 W216817 W216820 W216807 W216835 W216819 W216816 W216812 W216828
14235-672CS 845.09 USD InStock
14235-672 14235-650 14235-630 14235-674 14235-652 14235-670 14235-658 14235-614 14235-636 14235-616 14235-638 14235-632 14235-654 14235-676 14235-678 14235-612 14235-656 14235-634 14235-628 14235-660 14235-682 14235-640 14235-662 14235-680 14235-624 14235-646 14235-668 14235-648 14235-626 14235-620 14235-642 14235-666 14235-622 14235-644 14235-618
Boston Round Bottles, Clear, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton Boston round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
Boston round bottles are available with a choice of PTFE faced foam polyethylene liner with polypropylene cap, poly-vinyl liner with polypropylene cap, PE Cone liner with black phenolic cap, or rubber liner with black phenolic cap.
These narrow mouth bottles facilitate pouring and storing of liquids. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. Partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Available with caps attached or in bulk packs without caps. When included, caps come attached to bottles. Extra screw caps can be purchased separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty