Would you like to log in again?
Boston Round Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W216855 W216849 W216859 W216857 W216864 W216852 W216840 W216851 W216845 W216856 W216858 W216836 W216848 W216861 W216866 W216870 W216869 W216850 W216842 W216863 W216839 W216837 W216862 W216846 W216860 W216841 W216868 W216847 W216843 W216865 W216854 W216867 W216838 W216853 W216844
14235-694CS 134.43 USD InStock
14235-694 14235-696 14235-750 14235-690 14235-692 14235-734 14235-712 14235-714 14235-736 14235-698 14235-730 14235-752 14235-710 14235-732 14235-728 14235-706 14235-708 14235-740 14235-684 14235-702 14235-724 14235-746 14235-726 14235-704 14235-748 14235-742 14235-720 14235-686 14235-722 14235-744 14235-700 14235-688 14235-716 14235-738 14235-718
Boston Round Bottles, Amber, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton Boston round bottles are manufactured from amber soda-lime glass that conforms to USP Type III requirements.
Boston round bottles are offered with a choice of PTFE faced foam polyethylene liner with polypropylene cap, poly-vinyl liner with polypropylene cap, PE cone liner with black phenolic cap, or rubber liner with black phenolic cap.
These narrow mouth bottles facilitate pouring and storage of liquids. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. The partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required.
Ordering information: Bottles are available with caps attached or in bulk packs without caps. Extra screw caps can be purchased separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty