Would you like to log in again?
French Square Bottles, Clear, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W216875 W216882 W216887 W216894 W216871 W217881 W216900 W216901 W216877 W216890 W216881 W216878 W216880 W216889 W216893 W216902 W216892 W216876 W216883 W216899 W216896 W216872 W216873 W216895 W216885 W216884 W216879 W216874 W216897 W216886 W216888 W216898 W216891
14235-792CS 165.54 USD InStock
14235-792 14235-770 14235-772 14235-794 14235-790 14235-812 14235-778 14235-756 14235-814 14235-758 14235-774 14235-796 14235-810 14235-776 14235-754 14235-798 14235-808 14235-804 14235-806 14235-782 14235-760 14235-762 10847-500 14235-784 14235-780 14235-768 14235-800 14235-802 14235-786 14235-764 14235-766 14235-788 14235-816
French Square Bottles, Clear, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
French square bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
  • Square design allows for full utilization of any storage area
  • Maximizing space efficiency
Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. A partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labeling and/or sealing is required. Extra screw caps can be purchased separately.
Ordering information: Available with caps attached or bulk packs without caps. When included, caps are supplied attached to bottles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty