Would you like to log in again?
Valumetric™ Bottles, Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Graduated Valumetric™
W216993 W216977 W216973 W216970 W216991 W216990 W216979 W216976 W216986 W216985 W216975 W216987 W216980 W216967 W216988 W216992 W216968 W216984 W216966 W216974 W216971 W216972 W216969 W216982 W216978 W216989 W216981
14235-990CS 830.41 USD InStock
14235-990 14235-992 14235-970 14235-998 14235-954 14235-976 14235-956 14235-994 14235-972 14235-950 14235-952 14235-996 14235-974 14235-948 14235-980 14235-982 14235-960 14235-966 14235-988 14235-944 14235-946 14235-968 14235-984 14235-962 14235-964 14235-986 14235-958
Valumetric™ Bottles, Graduated, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
These clear glass bottles are ideal for use as sample bottles.
  • Graduated in ml and oz.
They are also useful for dry powders, soil samples, and specimen storage.
Ordering information: When included, caps come attached to bottles.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty