Would you like to log in again?
AC Round Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
W217004 W217000 W217005 W217001 W216996 W216998 W217002 W216997 W216999 W216995 W217008 W217007 W217009 W217006 W217003
14236-020CS 314.84 USD InStock
14236-020 14236-030 14236-002 14236-024 14236-012 14236-022 14236-010 14236-006 14236-028 14236-016 14236-004 14236-026 14236-014 14236-008 14236-018
AC Round Bottles, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Atlantic City round bottles are ideal for sampling use and can also be used to store dry powders, soil samples, and specimens.
  • Manufactured from Wheaton '800' glass
Certifications: Meets USP Type III standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty