Would you like to log in again?
BOOT ORANGE HS N/ESD S/3257-LG
Supplier: ALSICO USA
Boot^ With rigid tan sole. Wraparound strap and buckle closure at instep, color-coded for size identification. 19" high. Elastic top closure with adjustable snaps on tabs. One snap on back of leg to attach to coverall. Two snaps on tab, four snaps on hem. Color: Orange. Size: Large.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOOT ORANGE HS N/ESD S/3257-LG 14236-174 B39585SP2L Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty