Would you like to log in again?
COVERALL ORANGE S/3162-XL
Supplier: ALSICO USA
Coverall^ Long raglan sleeves with ESD polyester knit cuffs. Full length limited-linting front zipper fly closure with 2-ply fly stiffener. Military collar with color-coded hanger loop. Snap adjustments at collar and ankles. Additional snaps on back of legst. Color: Orange. Size: X-Large.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVERALL ORANGE S/3162-XL 14236-240 C39587SP2XL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty