Would you like to log in again?
Uvex Genesis XC™ Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
uvex genesis XC® Uvex Genesis XC®
S3304X S3307 S3309 S3308 S3300HS S3302 S3308 S3303X S3302 S3306HS S3306 S3301X S3301 S3305 S3305 S3301X S3301 S3300 S3309 S3301HS S3300 S3305
14236-383EA 34.71 USD InStock
CA14236-392 14236-383 CA14236-370 14236-371 CA14236-380 CA14236-390 14236-392 14236-381 14236-387 CA14236-384 14236-386 CA14236-362 CA14236-372 14236-363 14236-373 10790-628 CA14236-388 14236-389 CA14236-376 CA14236-386 14236-377 10790-656
Uvex Genesis XC™ Protective Eyewear, Honeywell Safety
Safety Glasses
Wraparound safety eyewear offers extended protection from particles and debris with more than 180° of peripheral coverage.
  • Multi-Material Technology® (MMT) molding provides all-day comfort
  • Ergonomically designed frame relieves pressure points
  • Lenses and cushioned temples can be adjusted for a more personalized fit
Several lens tints are available to suit most work applications. These include SCT Spectrum Control Technology® lenses that absorb select wavelengths of radiant energy, and Infra-dura® shades for welding, brazing, and cutting. The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® AF lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme™ lens coating features a combination of Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Certifications: ANSI Z87+ /CSA Z94.3
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty