Molded from a high-density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
CE marked.
The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The anterior printing area is at a 45° angle, offering an unobstructed view of the writing surface and making the cassettes compatible with automated labeling instruments. Biopsy cassettes are designed to hold biopsy samples securely during the embedding process. One-piece disposable plastic lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss.
Ordering information: Cassette bases are packaged in E-Z LOAD™ sleeves of 50 bases, secured by a holding tie. Each case contains two boxes of 20 sleeves.