Would you like to log in again?
Dip N Count® Urinary Culture Test, Starplex Scientific
Supplier: Starplex Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Dip N Count®
B30U0 B30U0
14276-276CS 229.4 USD InStock
14276-276 CA73250-022
Dip N Count® Urinary Culture Test, Starplex Scientific
Microbiology Test Systems Microorganism Tests
Dip N Count® is a semi-quantitative culture test for the detection of various types of bacteria that cause urinary tract infections
This Dip N Count® test provides paddles that combine two culture media types. MacConkey agar with Crystal Violet is a selective and differential medium that supports the growth of gram-negative bacteria and inhibits that of most gram-positive bacteria. C.L.E.D. agar is an electrolyte deficient medium that prevents the characteristic swarming of proteus. The inclusion of lactose allows for the differentiation of lactose fermenting bacteria by changing the color of the medium from pale green to yellow.
Tests should be stored at 4 to 25°C (39.2 to 77°F) for optimum shelf life. Each lot is tested to meet CLSI standards.
The test results can show the presence or absence of potential urinary pathogens, the approximate concentration of bacteria in CFU/mL or CFU/L, and a presumptive identification of the specific bacteria type.
Ordering information: Packaged 10 per box, 10 boxes per case. A quality control certificate is supplied for each lot.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty