Would you like to log in again?
Samco™ General-Purpose Transfer Pipettes, Thermo Scientific
Supplier: Thermo Scientific
204-20S 335-20S 202-20S 273-20S 204 202-10S 691 273 336 335-1S 336-1S 202 691-20S 273-1S 336-20S 204-1S 273 273-1S 202-1S 335-11
73990-092CS 1259.29 USD InStock
73990-092 14670-419 490014-708 14670-418 14670-417 73990-098 14670-147 73990-144 14670-003 14670-103 490014-700 14670-114 14670-454 73990-014 14670-001 73990-016 14670-149 490014-702 14670-468 14670-006
Samco™ General-Purpose Transfer Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Samco™ General-Purpose Transfer Pipettes feature a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving.
This also prevents cross-contamination. Transfer Pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface reduces loss of cells and valuable proteins due to binding. They can be frozen in liquid nitrogen, or heat-sealed for storage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty