Would you like to log in again?
Samco™ Sedi-Pet® Disposable Transfer Pipettes, Thermo Scientific
Supplier: Thermo Scientific
Sedi-Pet®
242 251-20S 242 252 251 241 251-1S
14670-170CS 1022.3 USD InStock
14670-170 14670-191 14670-180 14670-196 490014-722 14670-169 14670-345
Samco™ Sedi-Pet® Disposable Transfer Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Polyethylene pipettes are used for blood bank applications, such as sedimentation procedures.
The long flexible stem can be bent to draw liquids from narrow or small volume tubes into the bulb. Pipette stems are readily heat-sealed for conversion to liquid-tight storage and transport vessels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty