Would you like to log in again?
VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Supplier: VWR
VWR®
24672.608 24672.380 24672.200
14672-200CS 334.59 USD InStock
14672-200 14672-608 14672-380
VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Pipettes Pasteur/Transfer Pipettes
VWR® Disposable Pasteur Pipettes are ideally suited for use in both clinical and industrial laboratories.
  • Made up of borosilicate glass 7.0
  • Ideal for both clinical and industrial use
  • Supplied in convenient USE-Packs® packaging
Disposable Pasteur pipettes provide a convenient and time-efficient means for inoculating culture media and transferring samples and reagents in both clinical and industrial laboratories. No cotton plug. Pipette body Ext.Ø: 7 mm. Capillary stem Ext.Ø: approximately 1.5 mm. Controlled drop pipette (14672-608) delivers 22 to 28 drops per ml.
Packaging: Supplied in convenient USE-Packs® packages, each containing 200 pipettes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty