Would you like to log in again?
CR-1 Cleanroom Vacuum Cleaner System, Tiger-Vac®
Supplier: Tiger-Vac
Tiger-Vac®
110760A 110760A
14900-000EA 1616.04 USD InStock
14900-000 CA14900-000
CR-1 Cleanroom Vacuum Cleaner System, Tiger-Vac®
Vacuum Cleaners
Ideal for cleanroom and laboratory maintenance, this portable vacuum system features an ULPA (Ultra Low Penetration Air) filter with 99.999% efficiency on particles as small as 0.12µm
The vacuum's filtration system includes disposable paper filter bag with 7.6L (2gal.) recovery capacity and washable cloth main filter. System features a thru-flow motor with thermal protection. Compact size allows for easy handling and storage. Mounted on four swivel casters for maximum maneuverability.
Certifications: Compatible with ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions.
Ordering information: The Tiger Vac® CR-1 vacuum system includes ULPA filter, five paper filter bags, cloth main filter, 10m (33') power cord, 2.4m (8') suction hose, two straight steel extension wands, 36cm (14") floor tool, 20cm (8") crevice tool, 13cm (5") upholstery tool, and 8cm (3") dusting brush.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty