Would you like to log in again?
ComfortGrip® Latex Gloves, Microflex®, Ansell
Supplier: Ansell
ComfortGrip®
CFG-900-S CFG-900-L CFG-900-XS CFG-900-M CFG-900-S CFG-900-XS CFG-900-XL CFG-900-M CFG-900-XL CFG-900-L
14900-540CS 228.21 USD InStock
CA14900-540 14900-540 CA14900-532 14900-534 14900-532 14900-538 CA14900-538 14900-536 CA14900-534 CA14900-536
ComfortGrip® Latex Gloves, Microflex®, Ansell
Gloves
Disposable, medical-grade examination gloves are made of natural rubber latex, which offers excellent barrier integrity and elasticity for comfortable hand protection.
  • Powder-free natural rubber latex
  • Ambidextrous
  • Fully textured, beaded cuff
These low-protein gloves contain ≤50 µg of total water-extractable protein per gram.
Certifications: Gloves meet or exceed ASTM D3578 standards.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty