Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP Converter Plates, Analytik Jena
Supplier: Analytik Jena
38-0191-01 38-0191-02 38-0200-01 38-0191-04 38-0191-01 38-0191-02 38-0191-04 38-0200-01 38-0325-01
21474-892EA 749.67 USD InStock
CA21474-892 21474-892 CA15000-088 CA11500-064 CA82026-878 82026-878 15000-088 89237-310 95025-406
UVP Converter Plates, Analytik Jena
Transilluminators UV/White Light Transilluminators
UV to White Light Converter Plate measuring 21 x 26 cm, converts Analytik Jena transilluminator’s ultraviolet light to white light for viewing protein gels and Coomassie blue stained or silver stained media
Visi-Blue™ converter plate is designed to convert 302 nm UV light to 480 nm blue light for use with SYBR® Green, SYPRO® Orange and GFP stains. Surface is safety-fused into metal frame for durability.
Phosphored plexiglass uses a white diffuser to convert UV radiation. Plate housing is scratch-resistant metal. Handles on two sides allow easy transport.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty