Would you like to log in again?
Whatman™ Puradisc Syringe Filters, Nylon, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Puradisc™ Puradisc™
6799-2504 6789-1304 6768-1302 6789-1302 6789-1302 6752-2504 6789-1301 6790-0404 6750-2502 6790-1304 6753-2502 6790-1304 6790-1302 6751-2502 6789-0404 6750-2510 6789-0402 6753-2510 6753-2504 6790-0402 6750-2504 6789-1304 6751-2510 6753-2504 6768-1302 6799-2504 6753-2502 6768-1304 6753-2510 6768-1304 6751-2504 6751-2502 6789-1301 6752-2504 6751-2504 6790-1302 6790-0404 6750-2504 6751-2510 6750-2502 6786-0402 6750-2510 6790-0402 6799-2504
28137-552PK 2375.22 USD InStock
28137-552 28137-554 28205-512 CA10147-784 28137-550 28205-532 28205-510 CA11013-012 CA28137-548 28205-530 28137-616 CA28137-620 10192-068 CA28137-644 28137-614 CA28137-546 10148-000 10035-496 15000-254 TX6799-2504NAL CA28205-530 CA28205-510 CA28205-532 CA28205-512 CA11013-924 10035-498 CA28137-550 28137-620 CA28137-552 28205-502 28205-522 28205-500 CA11012-564 CA28137-554 28137-644 28137-546 28137-548 CA10148-000 CA28137-606 CA15000-254 CA28205-500 CA28205-522 CA28205-502 10035-500
Whatman™ Puradisc Syringe Filters, Nylon, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc syringe filters from Cytiva's business combine premium quality with economic efficiency. They are well suited for rapid, routine syringe filtration of samples up to 100 ml.
  • Eight different material options
  • Choice of 4, 13, and 30 mm syringe filter diameter to suit sample volume
  • Range of pore sizes for retaining a variety of particles
  • Optional sterile, medical-grade blister pack
  • Optional tube-tip format for accurate dispensing into a micro-vial
  • Recyclable after use
Puradisc syringe filters cater to a wide variety of sample types and applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation. Different Puradisc membrane media support diverse chemical solvents and compounds.
Puradisc is available in the following membrane materials: regenerated cellulose (RC), cellulose nitrate, nylon, polytetrafluoroethylene (PTFE), polyethersulfone (PES), polypropylene, polyvinylidene difluoride (PVDF), and cellulose acetate.
Cytiva's Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of applications. Cytiva also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for filtering larger volumes of harder-to-filter samples.
Nylon membrane filters are hydrophilic, flexible, tear-resistant, and autoclavable. They are resistant to a range of organic solvents and suitable for use with high pH samples. Nylon binds proteins, which makes it unsuitable for protein recovery applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty