Would you like to log in again?
Roto-Shake Genie™ 6–in–1 Multi-Purpose Rotator Rocker, Scientific Industries
Supplier: Scientific Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Roto-Shake Genie™
SI-1127 SI-1124 SI-1134 SI-1126 SI-1170 SI-1135 SI-1122 SI-1100 SI-1120 SI-1121 SI-1123 SI-1150 SI-1275 SI-1125 SI-1136 SI-1140 SI-1130 SI-1175
15001-002EA 235.75 USD InStock
15001-002 76808-692 15001-014 490015-482 89426-872 10753-556 15001-012 15001-006 14222-144 15001-004 15001-016 15001-008 89401-428 14222-140 14003-576 58815-176 15001-010 14003-580
Roto-Shake Genie™ 6–in–1 Multi-Purpose Rotator Rocker, Scientific Industries
Shakers and Mixers Rotating/Rocking Shakers
Space-saving multi-action platform rotator/rocker produces six different mixing motions: rotating, rotating/rocking, rotating/rolling, rolling/rocking, and rocking/shaking, as well as a combination of all of these.
  • Multi-action platform
  • Six mixing motions
  • Magnetized platform
The optional 3-D orbital shaker attachment transforms the unit into an orbital shaker
Clip plates hold similar or different tube sizes for various mixing motions, which can be achieved by varying the orientation of the clip plates on the magnetized platform. Ideal for use in cold rooms and low-temperature incubators. Complete and even soaking of membrane-based blot assays/washings. Easily cleaned and decontaminated. Rotator/rocker maintains set speed at temperatures from 0 to 38°C (32 to 100°F). Expansion kits are available for increased capacity and different containers.
The easily-removable, double-sided, stainless steel, magnetized platform converts from rotating to rocking with a simple mechanical adjustment. Platform can accommodate up to two 15.2x15.2cm (6x6") plastic bags per side, a 30.5x30.5cm (12x12") nonskid metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side.
Certifications: ETL listed to UL and CSA standards. CE marked.
Ordering information: Manufacturer’s two-year warranty. Unit supplied with 1 clip plate of each size (10–13mm tubes, 15–17mm tubes, and 28–30mm tubes), 1 nonskid metal rocker tray, and 4 magnetic stainless steel mounting strips. Expansion kit with 10–13 mm Tube Plates includes six plates with 12 clips per plate. Expansion kit with 15–17 mm Tube Plates includes six plates with six clips per plate. Expansion kit with 28–30 mm Tube Plates includes six plates with three clips per plate. Bag expansion kit includes two trays and 24 flexible magnetic mounting strips.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty