Would you like to log in again?
Urine Culture/Urinalysis Preservative Vial with 75 mg Boric Acid Tablet, Starplex Scientific
Supplier: Starplex Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
V102-CA V102-CA
15001-164CS 533.05 USD InStock
CA73240-130 15001-164
Urine Culture/Urinalysis Preservative Vial with 75 mg Boric Acid Tablet, Starplex Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Suitable for the transport of specimens for either Urinalysis or Urine Culture, this sterile, 10 mL Conical Vial utilizes a Boric Acid tablet to maintain the specimen for up to 72 hours
The vial is capped with a flat, blue cap which provides leak resistant transport. The vial has a patient identification label with fill line volume requirements. Studies available upon request.
Ordering information: Vials are packaged 500 per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty