Would you like to log in again?
Uvex Genesis® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
Genesis®
S3244 S3211X S3213X S3200 S3200HS S3210X S3260 S3240 S3241X S3204 S3240 S3260 S3243 S3200D S3200D S3244 S3211HS S3220X S3200 S3240X S3201X S3220 S3202 S3260X S3243 S3201HS S3241X S3203 S3200X S3202
14216-001EA 37.75 USD InStock
14216-001 15001-574 CA15001-582 15001-612 CA15001-620 15001-610 CA14231-228 15001-576 15001-616 CA15001-568 CA15001-622 CA14227-815 CA14227-817 94023-916 15001-570 CA14216-002 15001-584 CA14216-000 CA15001-610 15001-568 15001-622 CA15001-576 15001-620 CA15001-578 CA15001-616 CA94023-916 10790-626 10790-624 15001-580 14227-815
Uvex Genesis® Protective Eyewear, Honeywell Safety
Safety Glasses
Designed for comfort and compliance, Uvex Genesis® eyewear combines a lightweight, resilient multi-material frame with a soft elastomer brow guard for unsurpassed impact resistance.
  • Dual 9-base wraparound lens provides uncompromised peripheral vision and protection Elastomer brow-guard diffuses and deflects impact
  • Temple length and lens inclination are adjustable
  • Soft fingers conform to any nasal profile
  • Frames guaranteed for life
The dual 9-base wraparound polycarbonate lens provides uninterrupted peripheral coverage and visibility.
Clear lenses are appropriate for most indoor applications. Gold mirror, amber, and espresso lenses minimize the effects of sunlight and glare. The SCT-Reflect 50 lenses are designed for use in work applications with varying light conditions. This tint is a comfortable solution for workers who move between shaded or indoor locations and direct sunlight. SCT-Vermillion lenses are ideal for indoor applications requiring contrast, such as inspection. SCT-Blue lenses help counteract sodium vapor lighting. The Patriot® RWB frame features a red, white, and blue design with miniature U.S. flags at temples and 'United We Stand' inscribed on bottom right lens outside field of vision.
Soft, flexible 'fingers' adjust and conform to any nasal profile, minimizing slippage. A three-position ratchet lens inclination system completes the custom fit. A ventilation channel between frame and lens vents the eye socket area for outstanding moisture/fog control. Lens replacement is quick, easy, and economical. Ultra-dura® lens coating provides superior scratch and chemical resistance. Uvextreme® lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection. Dura-streme® lens coating features Ultra-dura® on the exterior and Uvextreme® on the interior for excellent protection and longevity.
Certifications: Eyewear meets military Vo ballistic test for impact and ANSI Z87+ and CSA Z94.3 standards.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty