Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Corrosive Storage Cabinets, SECURALL®
Supplier: Securall
C124 C330 C224 C160 C360 C345 C145 C130
15001-694EA 4253.25 USD InStock
15001-694 15001-688 15001-686 14226-894 14226-892 15001-692 14227-648 15001-690
Corrosive Storage Cabinets, SECURALL®
Furniture Storage Cabinets
Cabinet features adjustable shelving and leveling legs, three-point latching system, 5.1cm (2") vents with flash arrestors, flush-mounted locking handle, a sliding door, and a 5.1cm (2") leakproof sill
Caution: Not recommended for the storage of nitric or sulfuric acids.
Made with 18-gauge, galvanized steel with a double wall and a 3.8cm (11/2") air space. Shelves are equipped with removable corrosive-resistant polyethylene trays to contain spills and finished with two coats of corrosive-resistant blue epoxy enamel paint. Large warning label reads “CAUTION CORROSIVES.”
Certifications: Cabinets comply with OSHA and NFPA Code 30 requirements. All cabinets, with the exception of 14227-648, are FM approved.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty