Would you like to log in again?
Monoject™ Snap Caps, Covidien
Supplier: Covidien
1270019500 1270019404 1270019504 1270019505 1270019401 1270019502 1270019400 1270019400 1270019503
15070-042CS 1042.72 USD InStock
15070-042 15070-040 15070-052 15070-448 15070-446 15070-048 15070-104 CA15070-044 15070-044
Monoject™ Snap Caps, Covidien
Closures Test/Sample Tube Closures
Monoject snap caps are ideal for resealing most evacuated glass or plastic blood collection tubes
The caps feature a thumb tab for easy on and off.
Ordering information: These caps are for use with blood collection tubes only. Not for use with culture tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty