Would you like to log in again?
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Supplier: VWR
VWR®
K2600-VWR K2502-VWR K2508-VWR K2511-VWR
15070-296CS 86.24 USD InStock
15070-296 15070-294 15070-292 15070-290
VWR® Autoanalyzer Sample Cups, Polystyrene, Disposable
Sample Containers Sample Cups for Analyzers
These transparent polystyrene sample cups are resistant to most dilute aqueous solutions.
  • Compatible with most autoanalyzers
  • Wide base prevents spillage
Cups are available for use with Technicon®, Abbott®, Hitachi®, and most other autoanalyzers
Autoanalyzer sample cups are not autoclavable.
The wide-base design prevents tipping and spilling. The conical bottom is standard and recommended for microsampling.
Ordering information: The 2.0 mL cup (15070-290) also fits the Hyland laser-beam analyzer. Push-on polyethylene cap (15070-089) fits cups 15070-290, -293, and -294.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty