Would you like to log in again?
Ultrajet® 70 Duster, ITW Chemtronics®
Supplier: ITW Chemtronics
Ultrajet®
ES1015
15145-520CS 506.54 USD InStock
15145-520
Ultrajet® 70 Duster, ITW Chemtronics®
Sprays
This extra-strength duster removes dust, dirt, and other dry contaminants
Economizes solvent usage by removing layers of dry particulate first so the solvent can work immediately on more difficult-to-remove soils. Ideal for use on keyboards, circuit boards, surface mount devices, computers and other electronic components, assemblies, and photographic and video equipment. Nonflammable. Ozone-safe, CFC- and HCFC-free. 100% ultra-pure HFC-134a leaves no residues. Completely safe for use on most plastics.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty