Would you like to log in again?
Para-Pak® Stool Transport Systems, Meridian Bioscience
Supplier: Meridian Bioscience
Para-Pak®
300412 900112 900312 400112 300812 330284 300112 300212 900412 900612 301012
15157-014CS 2924.67 USD InStock
15157-014 15157-013 15157-024 15157-004 15157-010 15157-044 15157-022 15157-030 15157-018 15152-028 15157-006
Para-Pak® Stool Transport Systems, Meridian Bioscience
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
One-vial, two-vial, and three-vial systems are available
LV-PVA vials contain low viscosity polyvinyl alcohol. SAF vials contain sodium acetate–10% formalin.
Bulk poly bottles of LV-PVA for preservation and transportation and trichrome stain for the identification of parasites are also available.
C & S vials are intended for the transport of enteric pathogens and microbiological examination; all other vials are intended for the preservation and transportation of fecal samples for parasitology examination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty