Would you like to log in again?
Peel-A-Way® Disposable Embedding Molds, Polysciences
Supplier: Polysciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Peel-A-Way®
18986-1 18646A-1 18985-1 18646C-1 18646B-1
15160-099CS 395.14 USD InStock
15160-099 15160-157 15160-215 15160-339 15160-270
Peel-A-Way® Disposable Embedding Molds, Polysciences
Moulds Moulds for Histology
Disposable paraffin molds are designed to "Peel-A-Way" from the solidified embedded block
For plastic or paraffin embedding. Eliminates need for release agent, additional parts, and block trimming. Inside fingers hold ID slip, which becomes embedded in block for positive, permanent identification.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty