Would you like to log in again?
Nageotte Counting Chamber, Hausser Scientific
Supplier: Hausser Scientific
4000
15170-090EA 923.16 USD InStock
15170-090
Nageotte Counting Chamber, Hausser Scientific
Slides Counting Chambers
The Nageotte hemocytometer is designed to allow users to manually count low leukocyte concentrations, specifically those beyond the capabilities of a standard hemocytometer
The Nageotte chamber is supplied with two reusable coverslips (30x33x0.5mm) and 0.5oz. (approx. 35pcs.) of disposable coverslips (30x33x0.15mm).
Two gridded counting areas are subdivided into 40 rectangular areas. Each of the two gridded areas has a volume of 50µL, for a total volume of 100µL per Nageotte chamber. Accurate measurements can be obtained down to 0.1cell/µL (100cell/mL). Cell depth is 0.5mm. An H-shaped moat separates the two gridded areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty