Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Giemsa's stain, powder, HARLECO®, certified by the Biological Stain Commission for hematology, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

228-12 228-12
15204-122EA 511.69 USD InStock
CA15204-122 15204-122
Giemsa's stain, powder, HARLECO®, certified by the Biological Stain Commission for hematology, Sigma-Aldrich®
Giemsa's stain


Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty