Would you like to log in again?
MERIFLUOR® IFA Test Kits, Meridian Bioscience
Supplier: Meridian Bioscience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MERIFLUOR®
250050 222030
15205-012EA 1270.71 USD InStock
15205-012 15157-042
MERIFLUOR® IFA Test Kits, Meridian Bioscience
Clinical Diagnostic Supplies Clinical Reagents and Kits
IFA assays provide fluorescent interpretation based on monoclonal antibodies.
The Cryptosporidium/Giardia kit detects Cryptosporidium oocysts and Giardia cysts in fecal samples. It contains detection reagent, counter stain, wash buffer, positive and negative control, mounting medium, treated slides, and transfer loops to conduct 50 determinations.
The Pneumocystis kit detects Pneumocystis carinii cysts and prophozoites in upper respiratory infections. It contains detection reagent, mounting medium, slides, and transfer pipets to conduct 30 determinations.
CE marked.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty